Assay Details
Target Gene Details
Entrez Gene ID: | 214048 |
Gene Name: | La ribonucleoprotein domain family, member 1B |
Gene Aliases: |
1700108L22Rik, 4933421B21Rik, AA690246, Larp2 |
Location: |
Chr.3:40950631-40977797 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Larp1b | NM_001040399.1 | NP_001035489.1 | ||
| AK076732.1 | BAC36459.1 | |||
| AK160155.1 | BAE35661.1 | |||
| AK163348.1 | BAE37311.1 | |||
| BC025528.1 | AAH25528.1 | |||
| DQ553655.1 |
More Information
Set Membership: |
|

Genomic Map