Assay Details
Target Gene Details
Entrez Gene ID: | 28036 |
Gene Name: | La ribonucleoprotein domain family, member 7 |
Gene Aliases: |
C330027G06Rik, D3Wsu161e |
Location: |
Chr.3:127536714-127553349 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Larp7 | NM_138593.2 | NP_613059.2 | ||
| BC029178.1 | ||||
| BC132266.1 | ||||
| BC132268.1 | ||||
| BM946683.1 |
More Information
Set Membership: |
|

Genomic Map