Assay Details
Target Gene Details
Entrez Gene ID: | 381457 |
Gene Name: | cornulin |
Gene Aliases: |
Gm1015 |
Location: |
Chr.3:93144787-93149720 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crnn | NM_001081200.1 | NP_001074669.1 |
More Information
Set Membership: |
|

Genomic Map