Assay Details
Target Gene Details
Entrez Gene ID: | 74343 |
Gene Name: | CREB regulated transcription coactivator 2 |
Gene Aliases: |
4632407F12Rik, Torc2, mTORC2 |
Location: |
Chr.3:90254168-90264125 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crtc2 | NM_028881.2 | 10 | 993 | NP_083157.1 |
| AK014553.1 | 5 | 1364 | BAB29427.1 | |
| AK079382.1 | 8 | 1741 | ||
| AK156192.1 | 10 | 972 | BAE33618.1 | |
| AK169944.1 | 6 | 642 | BAE41473.1 | |
| AK172084.1 | BAE42818.1 | |||
| BC023091.1 | 6 | 583 | AAH23091.1 | |
| BC023831.1 | 10 | 988 | AAH23831.1 | |
| BC032183.1 | 10 | 998 | AAH32183.1 | |
| BC033300.1 | 10 | 988 | AAH33300.1 | |
| CV783724.1 | 7 | 497 |
More Information
Set Membership: |
|

Genomic Map