Assay Details
Target Gene Details
Entrez Gene ID: | 14658 |
Gene Name: | glycine receptor, beta subunit |
Gene Aliases: |
AI853901, Glyrb, spa, spastic |
Location: |
Chr.3:80843598-80913660 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glrb | NM_001281969.1 | NP_001268898.1 | ||
| NM_010298.6 | NP_034428.2 | |||
| AK082578.1 | ||||
| AK083251.1 | BAC38831.1 | |||
| AM420316.1 | ||||
| BC037605.1 | AAH37605.1 | |||
| U09399.1 | AAA61874.1 | |||
| X81202.1 | CAA57076.1 |
More Information
Set Membership: |
|

Genomic Map