Assay Details
Target Gene Details
Entrez Gene ID: | 14658 |
Gene Name: | glycine receptor, beta subunit |
Gene Aliases: |
AI853901, Glyrb, spa, spastic |
Location: |
Chr.3:80843598-80913660 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glrb | NM_001281969.1 | 5 | 496 | NP_001268898.1 |
| NM_010298.6 | 5 | 546 | NP_034428.2 | |
| AK082578.1 | 5 | 549 | ||
| AK083251.1 | 4 | 400 | BAC38831.1 | |
| AM420316.1 | 4 | 379 | ||
| BC037605.1 | 5 | 514 | AAH37605.1 | |
| U09399.1 | 5 | 455 | AAA61874.1 | |
| X81202.1 | 4 | 338 | CAA57076.1 | |
| X81203.1 | 2 | 121 | ||
| X81204.1 |
More Information
Set Membership: |
|

Genomic Map