Assay Details
Target Gene Details
Entrez Gene ID: | 14864 |
Gene Name: | glutathione S-transferase, mu 3 |
Gene Aliases: |
Fsc2, mGSTM5 |
Location: |
Chr.3:107963696-107969174 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gstm3 | NM_010359.2 | NP_034489.1 | ||
| BC044927.1 | AAH44927.1 | |||
| J03953.1 | AAA37748.1 |
More Information
Set Membership: |
|

Genomic Map