Assay Details
Target Gene Details
Entrez Gene ID: | 76893 |
Gene Name: | ceramide synthase 2 |
Gene Aliases: |
0610013I17Rik, AI225939, Lass2, TRH3 |
Location: |
Chr.3:95314805-95323588 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cers2 | NM_029789.1 | NP_084065.1 | ||
| AK002626.1 | ||||
| AK050161.1 | BAC34102.1 | |||
| AK146106.1 | ||||
| AK159325.1 | BAE34991.1 | |||
| AY029532.1 | AAK40300.1 | |||
| BC006847.1 | AAH06847.2 |

Genomic Map