Assay Details
Target Gene Details
Entrez Gene ID: | 12807 |
Gene Name: | Hermansky-Pudlak syndrome 3 homolog (human) |
Gene Aliases: |
coa |
Location: |
Chr.3:19995945-20035376 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hps3 | NM_001146323.1 | 13 | 2079 | NP_001139795.1 |
| NM_080634.4 | 13 | 2475 | NP_542365.3 | |
| AF393747.1 | 13 | 2464 | AAK84181.1 | |
| AF393780.1 | 13 | 2467 | AAL84109.1 | |
| AK034377.1 | 13 | 2446 | BAC28690.1 | |
| AK036663.1 | 13 | 2483 | BAC29524.1 | |
| AK155497.1 | 13 | 2442 | ||
| AK157953.1 | 13 | 2461 | ||
| AK169855.1 | 12 | 2547 | ||
| AK170581.1 | 13 | 1991 | BAE41891.1 | |
| AK171997.1 | 13 | 2067 | BAE42766.1 | |
| BC020105.1 | 3 | 574 | AAH20105.1 | |
| BC058235.1 | 13 | 2449 | AAH58235.1 |

Genomic Map