Assay Details
Target Gene Details
Entrez Gene ID: | 12508 |
Gene Name: | CD53 antigen |
Gene Aliases: |
AI323659, Ox-44, Tspan25 |
Location: |
Chr.3:106758861-106790149 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd53 | NM_007651.3 | NP_031677.1 | ||
| AA177409.1 | ||||
| AK156510.1 | BAE33740.1 | |||
| AK160326.1 | BAE35739.1 | |||
| BC021310.1 | AAH21310.1 | |||
| BC052905.1 | AAH52905.1 | |||
| X97227.1 | CAA65864.1 | |||
| Z16071.1 |

Genomic Map