Assay Details
Target Gene Details
Entrez Gene ID: | 12807 |
Gene Name: | Hermansky-Pudlak syndrome 3 homolog (human) |
Gene Aliases: |
coa |
Location: |
Chr.3:19995945-20035376 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hps3 | NM_001146323.1 | NP_001139795.1 | ||
| NM_001146324.1 | NP_001139796.1 | |||
| NM_080634.4 | NP_542365.3 | |||
| AF393747.1 | AAK84181.1 | |||
| AF393780.1 | AAL84109.1 | |||
| AK034377.1 | BAC28690.1 | |||
| AK036663.1 | BAC29524.1 | |||
| AK037829.1 | BAC29881.1 | |||
| AK049204.1 | BAC33608.1 | |||
| AK079896.1 | ||||
| AK086167.1 | ||||
| AK149359.1 | ||||
| AK155497.1 | ||||
| AK157953.1 | ||||
| AK166194.1 | ||||
| AK169855.1 | ||||
| AK170581.1 | BAE41891.1 | |||
| AK171997.1 | BAE42766.1 | |||
| AK180528.1 | ||||
| BB660133.1 | ||||
| BC058235.1 | AAH58235.1 |

Genomic Map