Assay Details
Target Gene Details
Entrez Gene ID: | 12807 |
Gene Name: | Hermansky-Pudlak syndrome 3 homolog (human) |
Gene Aliases: |
coa |
Location: |
Chr.3:19995945-20035376 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hps3 | NM_001146323.1 | 5 | 716 | NP_001139795.1 |
| NM_001146324.1 | 5 | 1112 | NP_001139796.1 | |
| NM_080634.4 | 5 | 1112 | NP_542365.3 | |
| AF393747.1 | 5 | 1101 | AAK84181.1 | |
| AF393780.1 | 5 | 1104 | AAL84109.1 | |
| AK034377.1 | 5 | 1083 | BAC28690.1 | |
| AK036663.1 | 5 | 1120 | BAC29524.1 | |
| AK037829.1 | 5 | 611 | BAC29881.1 | |
| AK049204.1 | 5 | 1105 | BAC33608.1 | |
| AK079896.1 | 5 | 1104 | ||
| AK086167.1 | 6 | 1228 | ||
| AK149359.1 | 5 | 1098 | ||
| AK155497.1 | 5 | 1079 | ||
| AK157953.1 | 5 | 1097 | ||
| AK166194.1 | 5 | 1098 | ||
| AK169855.1 | 4 | 1184 | ||
| AK170581.1 | 5 | 628 | BAE41891.1 | |
| AK171997.1 | 5 | 704 | BAE42766.1 | |
| AK180528.1 | 2 | 180 | ||
| BC058235.1 | 5 | 1086 | AAH58235.1 |

Genomic Map