Assay Details
Target Gene Details
Entrez Gene ID: | 229363 |
Gene Name: | guanine monophosphate synthetase |
Gene Aliases: |
AA591640, AI047208 |
Location: |
Chr.3:63964819-64021490 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gmps | NM_001033300.2 | NP_001028472.2 | ||
| AK049755.1 | BAE20667.1 | |||
| AK143197.1 | BAE25299.1 | |||
| AK146654.1 | BAE27334.1 | |||
| AK167857.1 | BAE39875.1 | |||
| AK168239.1 | BAE40189.1 | |||
| AK169043.1 | BAE40832.1 | |||
| AK169701.1 | BAE41314.1 | |||
| BC080685.1 | AAH80685.1 | |||
| BC138411.1 | ||||
| BC138412.1 |
More Information
Set Membership: |
|

Genomic Map