Assay Details
Target Gene Details
Entrez Gene ID: | 26879 |
Gene Name: | UDP-GalNAc:betaGlcNAc beta 1,3-galactosaminyltransferase, polypeptide 1 |
Gene Aliases: |
B3galt3, b3GT3 |
Location: |
Chr.3:69573919-69598960 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3galnt1 | NM_020026.4 | NP_064410.1 | ||
| AK088407.1 | BAC40336.1 | |||
| BC003835.1 | AAH03835.3 | |||
| BY278563.1 |
More Information
Set Membership: |
|

Genomic Map