Assay Details
Target Gene Details
Entrez Gene ID: | 57912 |
Gene Name: | CDC42 small effector 1 |
Gene Aliases: |
1300002M12Rik, AW558204, Cdcse1, SCIP1, Spec1 |
Location: |
Chr.3:95228424-95236424 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc42se1 | NM_001038708.3 | NP_001033797.1 | ||
| NM_172395.3 | NP_765983.1 | |||
| AK004859.1 | BAC25099.1 | |||
| AK019052.1 | BAC25576.1 | |||
| AK028573.1 | BAC26014.1 | |||
| BC060964.1 | AAH60964.1 | |||
| CF105339.1 |
Target Gene Details
Entrez Gene ID: | 56772 |
Gene Name: | myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 11 |
Gene Aliases: |
AI839562, Af1q, Zfp692 |
Location: |
Chr.3:95218544-95230459 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mllt11 |
More Information
Set Membership: |
|

Genomic Map