Assay Details
Target Gene Details
Entrez Gene ID: | 67713 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 19 |
Gene Aliases: |
1810055D05Rik, AA959924, Tim14 |
Location: |
Chr.3:34057280-34081354 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc19 | NM_001026211.2 | NP_001021382.1 | ||
| NM_001286972.1 | NP_001273901.1 | |||
| NM_001286973.1 | NP_001273902.1 | |||
| NM_026332.4 | NP_080608.3 | |||
| AK007866.1 | BAB25317.1 | |||
| AK008267.1 | BAB25565.1 | |||
| AK010204.1 | BAB26767.1 | |||
| AK012051.1 | BAB27993.1 | |||
| BC024335.1 | AAH24335.1 | |||
| BY149679.1 | ||||
| CA469052.1 | ||||
| CJ123658.1 | ||||
| EL962106.1 |

Genomic Map