Assay Details
Target Gene Details
Entrez Gene ID: | 110135 |
Gene Name: | fibrinogen beta chain |
Gene Aliases: |
2510049G14Rik |
Location: |
Chr.3:83042303-83049790 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgb | NM_181849.2 | NP_862897.1 | ||
| AF413205.1 | AAL02225.1 | |||
| AK011118.1 | ||||
| AK011920.1 | ||||
| AK146960.1 | BAE27567.1 | |||
| AK168626.1 | BAE40486.1 | |||
| BC031715.1 | AAH31715.1 |
More Information
Set Membership: |
|

Genomic Map