Assay Details
Target Gene Details
Entrez Gene ID: | 54124 |
Gene Name: | CDC28 protein kinase 1b |
Gene Aliases: |
2410005G18Rik, 2610005D03Rik, AA407784, Cks1, sid1334 |
Location: |
Chr.3:89415472-89418291 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cks1b | NM_016904.1 | NP_058600.1 | ||
| AB025409.1 | BAA84694.1 | |||
| AK004101.1 | BAB23169.1 | |||
| AK010414.2 | BAC25295.1 | |||
| AK011314.1 | BAB27540.1 | |||
| AK011782.1 | BAB27837.1 | |||
| BC076579.1 | AAH76579.1 |
More Information
Set Membership: |
|

Genomic Map