Assay Details
Target Gene Details
Entrez Gene ID: | 50911 |
Gene Name: | exosome component 9 |
Gene Aliases: |
PM/Scl-75, Pmscl1, RRP45, p5, p6 |
Location: |
Chr.3:36552606-36565727 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Exosc9 | NM_019393.2 | 7 | 751 | NP_062266.1 |
| AF152841.1 | 7 | 724 | AAF73218.1 | |
| AF152842.1 | 7 | 773 | AAF73219.1 | |
| AK010253.1 | 7 | 726 | ||
| AK011636.1 | 7 | 702 | BAB27749.1 | |
| AK076096.1 | 7 | 733 | ||
| AK083794.1 | 6 | 990 | ||
| BC005622.1 | 7 | 730 | AAH05622.1 | |
| BC052156.1 | 7 | 723 | AAH52156.1 | |
| BC158075.1 | 7 | 702 |
More Information
Set Membership: |
|

Genomic Map