Assay Details
Target Gene Details
Entrez Gene ID: | 99571 |
Gene Name: | fibrinogen gamma chain |
Gene Aliases: |
3010002H13Rik, AI256424 |
Location: |
Chr.3:83007896-83015049 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgg | NM_133862.1 | NP_598623.1 | ||
| AF413206.1 | AAL02226.1 | |||
| AK013887.1 | ||||
| AK149375.1 | BAE28843.1 | |||
| AK149448.1 | BAE28884.1 | |||
| BC019506.1 | AAH19506.1 | |||
| BC019828.1 | AAH19828.1 |
More Information
Set Membership: |
|

Genomic Map