Assay Details
Target Gene Details
Entrez Gene ID: | 14696 |
Gene Name: | guanine nucleotide binding protein (G protein), beta 4 |
Gene Aliases: |
6720453A21Rik, G(beta)4 |
Location: |
Chr.3:32580332-32616535 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnb4 | NM_013531.4 | NP_038559.2 | ||
| AF277893.1 | AAF82124.1 | |||
| AF501886.1 | AAM15922.1 | |||
| AK036816.1 | BAC29589.1 | |||
| AK098139.1 | ||||
| AK152227.1 | BAE31054.1 | |||
| AK155309.1 | BAE33181.1 | |||
| AK158965.1 | BAE34746.1 | |||
| AK165084.1 | 4 | 3385 | ||
| AK167416.1 | BAE39504.1 | |||
| AK168302.1 | BAE40243.1 | |||
| AK169752.1 | ||||
| AK172147.1 | ||||
| AK180015.1 | ||||
| AK187079.1 | ||||
| AK198808.1 | ||||
| AK201932.1 | ||||
| BC028753.1 | AAH28753.1 | |||
| M63658.1 | AAA37664.1 | |||
| M87286.1 | AAA37756.1 | |||
| S86124.1 | AAB21609.1 |
More Information
Set Membership: |
|

Genomic Map