Assay Details
Target Gene Details
Entrez Gene ID: | 18415 |
Gene Name: | heat shock protein 4 like |
Gene Aliases: |
94kDa, AI461691, APG-1, Osp94 |
Location: |
Chr.3:40744355-40796103 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hspa4l | NM_011020.3 | NP_035150.3 | ||
| AB001926.1 | BAA19468.1 | |||
| AK144225.1 | BAE25783.1 | |||
| AK163459.1 | BAE37350.1 | |||
| BC012712.1 | AAH12712.1 | |||
| BC057002.1 | AAH57002.1 | |||
| BC110662.1 | AAI10663.1 | |||
| D49482.1 | BAA08446.1 | |||
| U23921.1 | AAC52610.1 |

Genomic Map