Assay Details
Target Gene Details
Entrez Gene ID: | 18415 |
Gene Name: | heat shock protein 4 like |
Gene Aliases: |
94kDa, AI461691, APG-1, Osp94 |
Location: |
Chr.3:40744355-40796103 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hspa4l | NM_011020.3 | 16 | 2048 | NP_035150.3 |
| AB001926.1 | 16 | 2014 | BAA19468.1 | |
| AK033950.1 | 3 | 277 | BAC28524.1 | |
| AK144225.1 | 16 | 2085 | BAE25783.1 | |
| AK161429.1 | 3 | 194 | BAE36391.1 | |
| BC012712.1 | 16 | 2117 | AAH12712.1 | |
| BC057002.1 | 16 | 2048 | AAH57002.1 | |
| BC110662.1 | 16 | 2068 | AAI10663.1 | |
| D49482.1 | 16 | 2042 | BAA08446.1 | |
| U23921.1 | 16 | 2165 | AAC52610.1 |

Genomic Map