Assay Details
Target Gene Details
Entrez Gene ID: | 71492 |
Gene Name: | Bardet-Biedl syndrome 7 (human) |
Gene Aliases: |
8430406N16Rik |
Location: |
Chr.3:36573140-36613489 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bbs7 | NM_027810.3 | 11 | 1241 | NP_082086.2 |
| AF521645.1 | 11 | 1240 | AAO16027.1 | |
| AK050291.1 | 11 | 1239 | BAC34169.1 | |
| AK132661.1 | 11 | 1169 | BAE21287.1 | |
| BC031505.1 | 11 | 1191 | AAH31505.1 |

Genomic Map