Assay Details
Target Gene Details
Entrez Gene ID: | 70762 |
Gene Name: | doublecortin-like kinase 2 |
Gene Aliases: |
6330415M09Rik, AU044875, CL2, CLICK2, Click-II, Dcamkl2 |
Location: |
Chr.3:86786150-86920884 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dclk2 | NM_001195496.1 | NP_001182425.1 | ||
| NM_001195497.1 | NP_001182426.1 | |||
| NM_001195498.1 | NP_001182427.1 | |||
| NM_001195499.1 | NP_001182428.1 | |||
| NM_001195500.1 | NP_001182429.1 | |||
| NM_027539.5 | NP_081815.3 | |||
| AB198721.1 | BAE06836.1 | |||
| AK049126.1 | ||||
| AK049179.1 | BAC33590.1 | |||
| AK082633.1 | BAC38555.1 | |||
| AY968049.1 | ||||
| AY968050.1 | ||||
| AY968051.1 | ||||
| BC056921.1 | AAH56921.1 |
More Information
Set Membership: |
|

Genomic Map