Assay Details
Target Gene Details
Entrez Gene ID: | 14359 |
Gene Name: | fragile X mental retardation gene 1, autosomal homolog |
Gene Aliases: |
1110050J02Rik, 9530073J07Rik, AI851072, Fxr1h, Fxr1p |
Location: |
Chr.3:34020058-34069980 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fxr1 | NM_001113188.1 | 8 | 709 | NP_001106659.1 |
| NM_001113189.1 | 8 | 709 | NP_001106660.1 | |
| NM_008053.2 | 8 | 709 | NP_032079.1 | |
| AF124385.1 | 3 | 210 | AAD30211.1 | |
| BC019139.1 | 8 | 690 | AAH19139.1 | |
| X90875.1 | 8 | 688 | CAA62383.1 |
More Information
Set Membership: |
|

Genomic Map