Assay Details
Target Gene Details
Entrez Gene ID: | 229445 |
Gene Name: | cathepsin O |
Gene Aliases: |
A330105D01Rik, A430056C07, AI118514 |
Location: |
Chr.3:81932616-81956725 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctso | NM_177662.2 | NP_808330.1 | ||
| AK034490.1 | BAC28728.1 | |||
| AK049470.1 | BAC33765.1 | |||
| AK143683.1 | ||||
| AK165930.1 | BAE38466.1 | |||
| AK166103.1 | BAE38573.1 | |||
| BC044664.1 | AAH44664.1 | |||
| BC098219.1 | AAH98219.1 |
More Information
Set Membership: |
|

Genomic Map