Assay Details
Target Gene Details
Entrez Gene ID: | 99696 |
Gene Name: | ankyrin repeat domain 50 |
Gene Aliases: |
AI662170, E430012K20Rik, mKIAA1223, mKIAA3019 |
Location: |
Chr.3:38449261-38484861 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd50 | NM_001167883.1 | 2 | 224 | NP_001161355.1 |
| AK129484.1 | 2 | 209 | BAC98294.1 |
More Information
Set Membership: |
|

Genomic Map