Assay Details
Target Gene Details
Entrez Gene ID: | 13040 |
Gene Name: | cathepsin S |
Gene Aliases: |
- |
Location: |
Chr.3:95526786-95556405 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctss | NM_001267695.2 | NP_001254624.2 | ||
| NM_021281.3 | NP_067256.4 | |||
| AF038546.1 | AAB94925.1 | |||
| AJ002386.1 | CAA05360.1 | |||
| AK028366.1 | BAC25906.1 | |||
| AK152191.1 |
More Information
Set Membership: |
|

Genomic Map