Assay Details
Target Gene Details
Entrez Gene ID: | 13040 |
Gene Name: | cathepsin S |
Gene Aliases: |
- |
Location: |
Chr.3:95526786-95556405 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctss | NM_001267695.2 | 5 | 729 | NP_001254624.2 |
| NM_021281.3 | 5 | 726 | NP_067256.4 | |
| AF038546.1 | 5 | 668 | AAB94925.1 | |
| AJ002386.1 | 5 | 681 | CAA05360.1 | |
| AK028366.1 | 5 | 728 | BAC25906.1 | |
| AK150274.1 | 5 | 660 | BAE29430.1 | |
| AK150758.1 | 4 | 572 | BAE29827.1 | |
| AK150842.1 | 4 | 572 | BAE29900.1 | |
| AK152191.1 | 5 | 679 | ||
| AK152618.1 | 4 | 572 | BAE31361.1 | |
| AK153540.1 | 2 | 328 | BAE32077.1 | |
| BC002125.1 | 5 | 635 | AAH02125.1 | |
| Y18466.1 | 1 | 183 | CAA77184.1 |
More Information
Set Membership: |
|

Genomic Map