Assay Details
Target Gene Details
Entrez Gene ID: | 13559 |
Gene Name: | E2F transcription factor 5 |
Gene Aliases: |
AU024671, E2F-5 |
Location: |
Chr.3:14578602-14606309 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E2f5 | NM_007892.2 | 3 | 541 | NP_031918.2 |
| AK156760.1 | 3 | 460 | BAE33842.1 | |
| BC003220.1 | 3 | 541 | AAH03220.1 | |
| X86925.1 | 3 | 407 | CAA60508.1 |
More Information
Set Membership: |
|

Genomic Map