Assay Details
Target Gene Details
Entrez Gene ID: | 229488 |
Gene Name: | family with sequence similarity 160, member A1 |
Gene Aliases: |
9930021J17Rik |
Location: |
Chr.3:85660063-85845419 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam160a1 | NM_172682.3 | 10 | 3048 | NP_766270.2 |
| AK036881.1 | 7 | 2034 | BAC29618.1 | |
| AK076386.1 | 2 | 1903 | BAC36319.1 | |
| BC094511.1 | 8 | 2259 | AAH94511.1 |

Genomic Map