Assay Details
Target Gene Details
Entrez Gene ID: | 229488 |
Gene Name: | family with sequence similarity 160, member A1 |
Gene Aliases: |
9930021J17Rik |
Location: |
Chr.3:85660063-85845419 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam160a1 | NM_172682.3 | NP_766270.2 | ||
| AK036881.1 | BAC29618.1 | |||
| BC094511.1 | AAH94511.1 | |||
| DV651332.1 |

Genomic Map