Assay Details
Target Gene Details
Entrez Gene ID: | 67306 |
Gene Name: | zinc finger, C2HC-type containing 1A |
Gene Aliases: |
3110050N22Rik, AI790358, AU023959, Fam164a |
Location: |
Chr.3:7503415-7553848 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zc2hc1a | NM_173181.3 | NP_775273.1 | ||
| AK014202.1 | ||||
| AK083941.1 | BAC39075.1 | |||
| AK085187.1 | ||||
| AK170609.1 | BAE41909.1 |
More Information
Set Membership: |
|

Genomic Map