Assay Details
Target Gene Details
Entrez Gene ID: | 14468 |
Gene Name: | guanylate binding protein 2b |
Gene Aliases: |
Gbp-1, Gbp1, Mag-1, Mpa-1, Mpa1 |
Location: |
Chr.3:142594816-142619179 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gbp2b | NM_010259.2 | NP_034389.2 | ||
| AK077796.1 | ||||
| EF494422.1 |
More Information
Set Membership: |
|

Genomic Map