Assay Details
Target Gene Details
Entrez Gene ID: | 69585 |
Gene Name: | hemochromatosis type 2 (juvenile) (human homolog) |
Gene Aliases: |
2310035L15Rik, 5230400G09Rik, AI414844, AI789733, DL-M, HJV, Rgmc, hemojuvelin |
Location: |
Chr.3:96525163-96529216 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hfe2 | NM_027126.4 | NP_081402.3 | ||
| AJ557515.1 | CAD89720.1 | |||
| AK014082.1 | BAC25423.1 | |||
| BC022603.1 | AAH22603.1 |

Genomic Map