Assay Details
Target Gene Details
Entrez Gene ID: | 73284 |
Gene Name: | DNA-damage-inducible transcript 4-like |
Gene Aliases: |
1700037B15Rik, 1700108M02Rik, REDD2, RTP801L, Smhs1 |
Location: |
Chr.3:137623672-137628332 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddit4l | NM_030143.4 | 2 | 274 | NP_084419.2 |
| AF327512.1 | AAL39013.1 | |||
| AF335325.1 | 1 | 74 | AAL38425.1 | |
| AK006617.1 | 2 | 273 | BAB24676.1 | |
| AK018944.1 | 2 | 276 | BAB31490.1 | |
| AK053333.1 | 2 | 212 | BAC35350.1 | |
| AK075693.1 | 2 | 163 | ||
| BC038131.1 | 2 | 173 | AAH38131.1 | |
| BC147181.1 | 1 | 53 | ||
| BC147182.1 | 1 | 53 |

Genomic Map