Assay Details
Target Gene Details
Entrez Gene ID: | 14469 |
Gene Name: | guanylate binding protein 2 |
Gene Aliases: |
- |
Location: |
Chr.3:142620663-142638008 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gbp2 | NM_010260.1 | 8 | 1382 | NP_034390.1 |
| AF077007.1 | 8 | 1382 | AAC98287.1 | |
| AF109168.1 | 8 | 1335 | AAD39746.1 | |
| AJ007970.1 | 8 | 1375 | CAA07797.1 | |
| AK150138.1 | 8 | 1318 | BAE29334.1 | |
| AK150146.1 | 4 | 684 | BAE29341.1 | |
| BC011336.1 | 8 | 1371 | AAH11336.1 | |
| BC032882.1 | 8 | 1397 | AAH32882.1 | |
| CT010249.1 | 7 | 1212 | CAJ18457.1 |
More Information
Set Membership: |
|

Genomic Map