Assay Details
Target Gene Details
Entrez Gene ID: | 16194 |
Gene Name: | interleukin 6 receptor, alpha |
Gene Aliases: |
CD126, IL-6R, Il6r |
Location: |
Chr.3:89868829-89913492 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il6ra | NM_010559.2 | NP_034689.2 | ||
| AK046337.1 | ||||
| AK082270.1 | ||||
| AK141197.1 | BAE24580.1 | |||
| X51975.1 | CAA36237.1 | |||
| X53802.1 | CAA37810.1 |
More Information
Set Membership: |
|

Genomic Map