Assay Details
Target Gene Details
Entrez Gene ID: | 16196 |
Gene Name: | interleukin 7 |
Gene Aliases: |
A630026I06Rik, Il-7, hlb368 |
Location: |
Chr.3:7572028-7613427 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il7 | NM_008371.4 | NP_032397.1 | ||
| AK040399.1 | ||||
| AK041307.1 | BAC30899.1 | |||
| AK042009.1 | ||||
| AK052452.1 | ||||
| AK139858.1 | BAE24162.1 | |||
| AK145184.1 | BAE26279.1 | |||
| BC110553.2 | AAI10554.1 | |||
| X07962.1 | CAA30779.1 |

Genomic Map