Assay Details
Target Gene Details
Entrez Gene ID: | 16497 |
Gene Name: | potassium voltage-gated channel, shaker-related subfamily, beta member 1 |
Gene Aliases: |
Akr8a8, Kvbeta1.1, mKv(beta)1 |
Location: |
Chr.3:64949049-65378225 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnab1 | NM_001289450.1 | NP_001276379.1 | ||
| NM_010597.4 | NP_034727.3 | |||
| AF033003.1 | AAB87085.1 | |||
| AK004666.1 | ||||
| AK015412.1 | BAB29837.1 | |||
| AK031362.1 | BAC27367.1 | |||
| AK138467.1 | BAE23670.1 | |||
| AK153200.1 | BAE31800.1 | |||
| BC014701.1 | AAH14701.1 | |||
| U65591.1 | AAB37262.1 | |||
| X97281.1 | CAA65936.1 |
More Information
Set Membership: |
|

Genomic Map