Assay Details
Target Gene Details
Entrez Gene ID: | 229320 |
Gene Name: | clarin 1 |
Gene Aliases: |
A130002D11Rik, USH3, Ush3a |
Location: |
Chr.3:58844028-58885212 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clrn1 | NM_153384.3 | 3 | 608 | NP_700433.1 |
| NM_153385.3 | 2 | 554 | NP_700434.1 | |
| NM_153386.3 | NP_700435.1 | |||
| AF495718.1 | 3 | 608 | AAM88775.1 | |
| AF495719.1 | 2 | 554 | AAM88776.1 | |
| AF495720.1 | AAM88777.1 | |||
| AK037267.1 | 3 | 272 | BAC29781.1 | |
| AK041623.1 | BAC31009.1 | |||
| AK042523.1 | BAC31282.1 | |||
| BC132254.1 | ||||
| BC132256.1 | ||||
| BC145202.1 | 2 | 536 |

Genomic Map