Assay Details
Target Gene Details
Entrez Gene ID: | 13638 |
Gene Name: | ephrin A3 |
Gene Aliases: |
AW494418, EFL-2, Ehk1-L, Epl3, LERK-3 |
Location: |
Chr.3:89313899-89322960 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Efna3 | NM_010108.1 | NP_034238.1 | ||
| BC107002.2 | ||||
| BC125003.1 |
More Information
Set Membership: |
|

Genomic Map