Assay Details
Target Gene Details
Entrez Gene ID: | 55932 |
Gene Name: | guanylate binding protein 3 |
Gene Aliases: |
AW228655, GBP-3, GBP-4, Gbp4 |
Location: |
Chr.3:142560016-142573213 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gbp3 | NM_001289492.1 | NP_001276421.1 | ||
| NM_001289493.1 | NP_001276422.1 | |||
| NM_018734.3 | NP_061204.3 | |||
| AK132266.1 | ||||
| BC019195.1 | AAH19195.1 | |||
| CK389037.1 | ||||
| U44731.1 | AAA86645.1 |
More Information
Set Membership: |
|

Genomic Map