Assay Details
Target Gene Details
Entrez Gene ID: | 55932 |
Gene Name: | guanylate binding protein 3 |
Gene Aliases: |
AW228655, GBP-3, GBP-4, Gbp4 |
Location: |
Chr.3:142560016-142573213 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gbp3 | NM_001289492.1 | 7 | 1160 | NP_001276421.1 |
| NM_001289493.1 | 6 | 985 | NP_001276422.1 | |
| NM_018734.3 | 6 | 1022 | NP_061204.3 | |
| AK132266.1 | 6 | 1021 | ||
| BC019195.1 | 7 | 1071 | AAH19195.1 | |
| U44731.1 | 6 | 874 | AAA86645.1 |
More Information
Set Membership: |
|

Genomic Map