Assay Details
Target Gene Details
Entrez Gene ID: | 68183 |
Gene Name: | breast carcinoma amplified sequence 2 |
Gene Aliases: |
6430539P16Rik, AI132645, C76366, C80030 |
Location: |
Chr.3:103171660-103179163 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcas2 | NM_026602.3 | 4 | 439 | NP_080878.2 |
| AA544668.1 | 4 | 439 | ||
| AK005825.1 | 4 | 405 | BAB24258.1 | |
| AK018786.1 | 4 | 406 | BAB31409.1 | |
| AK020098.1 | 4 | 402 | BAB31992.1 | |
| AK153513.1 | 4 | 402 | BAE32056.1 | |
| AK166967.1 | 4 | 402 | BAE39150.1 | |
| AY043239.1 | 4 | 391 | AAK85306.1 | |
| BC023382.1 | 4 | 419 | ||
| BC037062.1 | 4 | 422 | AAH37062.1 |
More Information
Set Membership: |
|

Genomic Map