Assay Details
Target Gene Details
Entrez Gene ID: | 241943 |
Gene Name: | coiled-coil domain containing 144B |
Gene Aliases: |
- |
Location: |
Chr.3:36007247-36053547 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc144b | NM_178418.4 | 1 | 57 | NP_848505.2 |
| AK133178.1 | ||||
| BC050811.1 | 1 | 59 | AAH50811.1 |

Genomic Map