Assay Details
Target Gene Details
Entrez Gene ID: | 73852 |
Gene Name: | DNA segment, Chr 3, ERATO Doi 751, expressed |
Gene Aliases: |
2810009O15Rik, 4930415G15Rik |
Location: |
Chr.3:41742606-41803292 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| D3Ertd751e | NM_001099785.1 | NP_001093255.1 | ||
| NM_001291048.1 | NP_001277977.1 | |||
| NM_027271.1 | NP_081547.1 | |||
| NM_028667.3 | NP_082943.2 | |||
| AK039725.1 | ||||
| AK045813.1 | BAC32501.1 | |||
| AK085404.1 | BAC39441.1 | |||
| AK138405.1 | ||||
| AK144064.1 |

Genomic Map