Assay Details
Target Gene Details
Entrez Gene ID: | 56790 |
Gene Name: | suppressor of Ty 20 |
Gene Aliases: |
AA667204, AI450544, D3Ertd300e, Fam48a, Supt20h, p38IP |
Location: |
Chr.3:54692761-54728763 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Supt20 | NM_019995.3 | 19 | 1682 | NP_064379.1 |
| AF139179.1 | 19 | 1686 | AAF61447.1 | |
| AK171787.1 | BAE42664.1 | |||
| AK208103.1 | 2 | 169 | ||
| AK208868.1 | 2 | 169 | ||
| AK210834.1 | 2 | 169 | ||
| BC052702.1 | 19 | 1671 | AAH52702.1 |
More Information
Set Membership: |
|

Genomic Map