Assay Details
Target Gene Details
Entrez Gene ID: | 56790 |
Gene Name: | suppressor of Ty 20 |
Gene Aliases: |
AA667204, AI450544, D3Ertd300e, Fam48a, Supt20h, p38IP |
Location: |
Chr.3:54692761-54728763 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Supt20 | NM_019995.3 | NP_064379.1 | ||
| AF139179.1 | AAF61447.1 | |||
| AK050619.1 | BAC34346.1 | |||
| AK083379.1 | BAC38893.1 | |||
| AK171787.1 | BAE42664.1 | |||
| AK188434.1 | ||||
| AK210270.1 | ||||
| BC052702.1 | AAH52702.1 |
More Information
Set Membership: |
|

Genomic Map